Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cantharidic Acid

Catalog No. 21015010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-015-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-015-010MG Supplier MilliporeSigma™ Supplier No. 21015010MG
Add to Cart
Add to Cart

A potent inhibitor of protein phosphatase 2A (IC50 = 50 nM).

A potent inhibitor of protein phosphatase 2A (IC50 = 50 nM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >95% by TLC
Target Proteins PP2A
CAS 28874-45-5
Molecular Formula C10H14O5
Formula Weight 214.2g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.