Learn More
Description
Specifications
Specifications
| Color | White |
| Description | 99% by titration |
| Product Type | Agonist |
| Target Proteins | Agonist for nicotinic and muscarinic acetylcholine receptors |
| CAS | 51-83-2 |
| Molecular Formula | C6H15ClN2O2 |
| Formula Weight | 182.7g/mol |
| Solubility | H2O, EtOH, or MeOH |
| Content And Storage | Hygroscopic |
| Form | Crystalline Solid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.