Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Carbonyl Cyanide m-Chlorophenylhydrazone

Catalog No. 215911250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-591-1250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-591-1250MG Supplier MilliporeSigma™ Supplier No. 215911250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protonophore. Uncoupling agent for oxidative phosphorylation that inhibits mitochondrial function. Approximately 100 times more effective than 2,4-dinitrophenol.

Protonophore. Uncoupling agent for oxidative phosphorylation that inhibits mitochondrial function. Approximately 100 times more effective than 2,4-dinitrophenol. Binds with cytochrome c oxidase with high affinity (Kd = 270 nM). Inhibits transport processes and depresses growth.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Target Proteins cytochrome c oxydase
CAS 555-60-2
Formula Weight 204.6g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.