Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Carboplatin

Catalog No. 21610025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-610-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-610-025MG Supplier MilliporeSigma™ Supplier No. 21610025MG
Add to Cart
Add to Cart

An analog of Cisplatin with reduced nephrotoxicity. Displays potent antitumor activity.

An analog of Cisplatin (Cat. No. 232120) with reduced nephrotoxicity. Displays potent antitumor activity.

Note: Athough soluble in DMSO, Carboplatin is reported to be rendered inactive due to ligand displacement by the nucleophilic sulfur of DMSO. Water with or without 5% glucose is recommended for solubilization prior to culture treatment.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Potent antitumor activity
CAS 41575-94-4
Molecular Formula C6H12N2O4Pt
Formula Weight 371.3g/mol
Solubility H2O with 5% Glucose
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.