Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Casein Kinase II Inhibitor III, TBCA

Catalog No. 2187105MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-871-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-871-05MG Supplier MilliporeSigma™ Supplier No. 2187105MG
Add to Cart
Add to Cart

The Casein Kinase II Inhibitor III, TBCA, also referenced under CAS 934358-00-6, controls the biological activity of Casein Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins Casein Kinase II
CAS 934358-00-6
Molecular Formula C9H4Br4O2
Formula Weight 463.8g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.