Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-1 Substrate VI, Fluorogenic

Catalog No. 6882255MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-822-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-822-55MG Supplier MilliporeSigma™ Supplier No. 6882255MG
Add to Cart
Add to Cart

Fluorogenic caspase-1 substrate.

Peptide Sequence: Ac-Tyr-Val-Ala-Asp-AFC
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Description 98±1% by HPLC
Target Proteins Fluorogenic caspase-1 substrate
Molecular Formula C33H36F3N5O10
Formula Weight 719g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.