Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-3/7 Inhibitor II

Catalog No. 2188321MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-883-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-883-21MG Supplier MilliporeSigma™ Supplier No. 2188321MG
Add to Cart
Add to Cart

The Caspase-3/7 Inhibitor II, also referenced under CAS 775289-20-8, controls the biological activity of Caspase-3/7. This small molecule/inhibitor is primarily used for Cancer applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Asp-Asn-Leu-Asp-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Single main spot with additional trace spot by TLC
Target Proteins caspase-3, caspase-7
CAS 775289-20-8
Molecular Formula C20H31N5O10
Formula Weight 501.5g/mol
Solubility DMSO or H2O
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.