Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-3 Inhibitor I, Cell-Permeable

Catalog No. 2354231MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-542-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-542-31MG Supplier MilliporeSigma™ Supplier No. 2354231MG
Add to Cart
Add to Cart

The Caspase-3 Inhibitor I, Cell-Permeable controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

  • Peptide Sequence: Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Glu-Val-Asp-CHO
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 97% by HPLC
Target Proteins caspase-3
Molecular Formula C94H158N20O27
Formula Weight 2000.4g/mol
Solubility DMSO
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.