Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-3 Inhibitor III

Catalog No. 2187501MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

The Caspase-3 Inhibitor III, also referenced under CAS 285570-60-7, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.

  • Peptide Sequence: Ac-Asp-Glu-Val-Asp-CMK
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white to Brown
Target Proteins caspase-3, caspase-6, caspase-7, caspase-8, caspase-10
CAS 285570-60-7
Molecular Formula C21H31ClN4O11
Formula Weight 551g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.