Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-6 Substrate III, Fluorogenic

Catalog No. 2187635MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-876-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-876-35MG Supplier MilliporeSigma™ Supplier No. 2187635MG
Add to Cart
Add to Cart

A fluorogenic substrate for caspase-6 (Mch2).

Peptide Sequence: Z-Val-Glu-Ile-Asp-AFC
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins A fluorogenic substrate for caspase-6 (Mch2
Molecular Formula C38H44F3N5O12
Formula Weight 819.8g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.