Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Inhibitor I, Biotin Conjugate

Catalog No. 2187421MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-874-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-874-21MG Supplier MilliporeSigma™ Supplier No. 2187421MG
Add to Cart
Add to Cart

The Caspase Inhibitor I, Biotin Conjugate controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

Peptide Sequence: Biotin-X-Val-Ala-Asp(OMe)-CH2F* (X = linker)
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description single spot by TLC
Target Proteins Caspase
Molecular Formula C30H49FN6O8S
Formula Weight 672.8g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.