Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Inhibitor II

Catalog No. 2187351MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-873-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-873-51MG Supplier MilliporeSigma™ Supplier No. 2187351MG
Add to Cart
Add to Cart

The Caspase Inhibitor II, also referenced under CAS 147837-52-3, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.

Peptide Sequence: Ac-Val-Ala-Asp-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins pan-caspase
CAS 147837-52-3
Molecular Formula C14H23N3O6
Formula Weight 329.4g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.