Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Inhibitor Set I

Catalog No. 2354291SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-542-91SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-542-91SET Supplier MilliporeSigma™ Supplier No. 2354291SET
Add to Cart
Add to Cart

The Caspase Inhibitor Set I controls the biological activity of Caspase-1. This collection of small molecule/inhibitor is primarily used for Cancer applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins caspase-1
Content And Storage Protect from moisture
Form Solid
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.