Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase Modulator I, 1541

Catalog No. 26415710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-415-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-415-710MG Supplier MilliporeSigma™ Supplier No. 26415710MG
Add to Cart
Add to Cart

A cell-permeable phenylimidazopyridinyl-methoxy coumarin compound that inhibits caspase-3 and caspase-6 activities (IC50 = 35 and 38μM, respectively) by targeting both the large and small subunit active si

A cell-permeable phenylimidazopyridinyl-methoxy coumarin compound that inhibits caspase-3 and caspase-6 activities (IC50 = 35 and 38μM, respectively) by targeting both the large and small subunit active sites. Kinetic studies with 1541 and its 8-Hydroxy analog (Cat. No. 529663) indicate that at suboptimal inhibitor : proenzyme molar ratio (100:1 or less), the compound does not effectively occupies both active sites of and stabilizes the proenzyme in an “on-state” conformation that facilitates the subsequent autoproteolysis or autoactivation process via the unoccupied active site of procaspase-3 and -6 (EC50 = 2.4 and 2.8μM, respectively). Shown to effectively induce apoptosis (up to 50μM) in various cancer cell lines (IC50 ranges from 3.8 to 8.7μM) in a caspase-3-dependent, but p53-, caspase-8-, and Bak-independent manner. Exhibits little inhibitory effect against active caspase-7 (30nM) even at concentrations as high as 50μM, nor does it activate procaspase-1 or -7 even after prolonged incubations upto 4 and 24 h, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Molecular Formula C24H17N3O4
Formula Weight 411.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.