Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Castanospermine, Castanospermum australe

Catalog No. 2187751MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-877-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-877-51MG Supplier MilliporeSigma™ Supplier No. 2187751MG
Add to Cart
Add to Cart

Plant alkaloid inhibitor of several β-glucosidases and α-glucosidases, including those involved in N-linked processing of glycoproteins.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins β-glucosidases
CAS 79831-76-8
Molecular Formula C8H15NO4
Formula Weight 189.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.