Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Catenin/Tcf Inhibitor II, PKF118-310

Catalog No. 21933110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-933-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-933-110MG Supplier MilliporeSigma™ Supplier No. 21933110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The β-Catenin/Tcf Inhibitor II, PKF118-310 controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Cancer applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Yellow
Description 95% by HPLC
Molecular Formula C7H7N5O2
Formula Weight 193.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.