Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Catenin/Tcf Inhibitor VI, ICG-001

Catalog No. 5047120001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-471-20001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-471-20001 Supplier MilliporeSigma™ Supplier No. 5047120001
Add to Cart
Add to Cart

A cell-permeable pyrazinopyrimidinecarboxamide that competes against β-catenin for CBP (CREB-binding protein) binding and selectively prevents CBP- but not p300-, dependent TCF/β-catenin transcriptional regulations both in cultures in vitro (5 to 25μM) and in mice in vivo (5mg/kg/day via osmotic pump infusion), while not affecting CBP-dependent transcriptional activities mediated by the AP-1 or CRE c

A cell-permeable pyrazinopyrimidinecarboxamide that competes against β-catenin for CBP (CREB-binding protein) binding and selectively prevents CBP- but not p300-, dependent TCF/β-catenin transcriptional regulations both in cultures in vitro (5 to 25μM) and in mice in vivo (5mg/kg/day via osmotic pump infusion), while not affecting CBP-dependent transcriptional activities mediated by the AP-1 or CRE complex. Shown to reactivate p300//TCF/β-catenin-mediated neuronal differentiation upon NGF stimulation in PS-1 L286V mutant-expressing PC-12 cultures and be efficacious in alleviating bleomycin- (0,08 units/50μL/mouse; administered intranasally; Cat. No. 203401) induced pulmonary fibrosis in mice.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins CREB-binding protein
Molecular Formula C33H32N4O4
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.