Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cathepsin G Substrate IV, Fluorogenic

Catalog No. 2194741MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-947-41MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-947-41MG Supplier MilliporeSigma™ Supplier No. 2194741MG
Add to Cart
Add to Cart

An internally quenched fluorogenic peptide substrate (Mca/Anb as fluorophore/quencher) that is preferentially cleaved by cathepsin G between Gnf and Ser with high efficiency and selectivity.

  • Packaged Under Inert Gas
  • Peptide Sequence: Mca-Phe-Val-Thr-Gnf∼Ser-Trp-Anb-NH2
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt
Description 97% by HPLC
Molecular Formula C61H67N13O15
Formula Weight 1222.3g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.