Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cathepsin K Inhibitor I

Catalog No. 2193775MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-937-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-937-75MG Supplier MilliporeSigma™ Supplier No. 2193775MG
Add to Cart
Add to Cart

The Cathepsin K Inhibitor I controls the biological activity of Cathepsin K. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Cbz-Leu-NH-CH2-CO-CH2-NH-Leu-Cbz
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins cathepsin K
Molecular Formula C31H42N4O7
Formula Weight 582.7g/mol
Solubility DMSO or MeOH
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.