Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cathepsin K Substrate II, Fluorogenic

Catalog No. 2195551MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-955-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-955-51MG Supplier MilliporeSigma™ Supplier No. 2195551MG
Add to Cart
Add to Cart

An internally quenched, highly cathepsin K-selective, fluorogenic peptide substrate that is efficiently cleaved by CatK (

An internally quenched, highly cathepsin K-selective, fluorogenic peptide substrate that is efficiently cleaved by CatK ( kcat/Km = 0.426μM-1 S-1), but not by Cathepsins B/L/S/F/H/V. Studies using WT and CatK-/- mice reveal that the substrate is not cleaved by other proteolytic enzymes present in fibroblast lysates.
TRUSTED_SUSTAINABILITY

Specifications

Color Bright Yellow
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Molecular Formula C40H50N14O12
Formula Weight 918.4g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from Light, Protect from Moisture
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.