Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cathepsin L Inhibitor, CAA0225

Catalog No. 2195021MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-950-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-950-21MG Supplier MilliporeSigma™ Supplier No. 2195021MG
Add to Cart
Add to Cart

An epoxysuccinyl peptide highly selective for rat liver cathepsin L (IC50

An epoxysuccinyl peptide highly selective for rat liver cathepsin L (IC50 = 1.9nM), while exhibiting little to no effect on rat liver cathepsin B at concentrations higher than 1000-5000 nM, in vitro . During autophagy, it has been shown through immunoblotting that CAA0225 (10μg/ml) effectively inhibits the degradation of LC3-II and GABARAP compared to control, in nutrient-deprived HeLa and Huh-7 cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Molecular Formula C28H29N3O5
Formula Weight 487.55g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.