Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caveolin-1 Scaffolding Domain Peptide, Cell-permeable

Catalog No. 2194821MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-948-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-948-21MG Supplier MilliporeSigma™ Supplier No. 2194821MG
Add to Cart
Add to Cart

Caveolin-1 Scaffolding Domain Peptide, Cell-permeable, blocks eNOS activity and cellular NO release in vitro and reduces inflammation and tumorigenesis in vivo.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Asp-Gly-Ile-Trp-Lys-Ala-Ser-Phe-Thr-Thr-Phe-Thr-Val-Thr-Lys-Tyr-Trp-Phe-Tyr-Arg-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins eNOS
Molecular Formula C228H335N61O49S
Formula Weight 4746.6g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.