Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CBFβ-Runx1 Inhibitor II, Ro5-3335

Catalog No. 21950610MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-950-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-950-610MG Supplier MilliporeSigma™ Supplier No. 21950610MG
Add to Cart
Add to Cart

A cell-permeable benzodiazepine that suppresses CBFα/Runx1-CBFβ transactivation activity (25μM; 293-0 cells) via direct bindings to both subunits of the heterodimeric transcription factor complex

A cell-permeable benzodiazepine that suppresses CBFα/Runx1-CBFβ transactivation activity (25μM; 293-0 cells) via direct bindings to both subunits of the heterodimeric transcription factor complex. Shown to inhibit oncogenic fusion CBFB-MYH11 -, TEL-RUNX1 -, and RUNX1-ETO -, dependent leukemia cells proliferation (IC50 = 1.1 to 21.7μM) in vitro and more effectively reduce peripheral blood c-kit+ population (300mg/kg/d via animal feed) than Cytarabine (Cat. No. 251010) in a murine Cbfb-MYH11 leukemia model in vivo . Comparing to CBFβ-Runx1 Inhibitor I (Cat. No. 219505), Ro5-3335 modulates the Runx1-CBFβ heterodimer formation without completely disrupting the subunits interaction.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins RUNX1 and CBFβ
Molecular Formula C13H10ClN3O
Formula Weight 259.69g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.