Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CBP-CREB Interaction Inhibitor

Catalog No. 217505250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-750-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-750-5250MG Supplier MilliporeSigma™ Supplier No. 217505250MG
Add to Cart
Add to Cart

The CBP-CREB Interaction Inhibitor, also referenced under CAS 92-78-4, controls the biological activity of CBP-CREB.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Description 97% by HPLC
CAS 92-78-4
Molecular Formula C17H12ClNO2
Formula Weight 297.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.