Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CDC25 Phosphatase Inhibitor I, BN82002

Catalog No. 2176915MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-769-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-769-15MG Supplier MilliporeSigma™ Supplier No. 2176915MG
Add to Cart
Add to Cart

The CDC25 Phosphatase Inhibitor I, BN82002, also referenced under CAS 396073-89-5, controls the biological activity of CDC25 Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation and Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description 95% by HPLC
Target Proteins CDC25A
CAS 396073-89-5
Molecular Formula C19H25N3O4
Formula Weight 359.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.