Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CDC42 Inhibitor III, ZCL278

Catalog No. 5005030001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-050-30001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-050-30001 Supplier MilliporeSigma™ Supplier No. 5005030001
Add to Cart
Add to Cart

A cell-permeable, non-cytotoxic, aqueous soluble (181μM in PBS, pH 7

A cell-permeable, non-cytotoxic, aqueous soluble (181μM in PBS, pH 7.4) pyrimidinylthiourea that targets Cdc42 GEF ITSN/intersectin and adjacent GTP/GDP binding site and effectively inhibits EGF-stimulated Cdc42 activity (1 h pretreatment; 50μM) and Cdc42-dependent microspike/filopodia formation in serum-starved Swiss 3T3 fibroblast cultures without affecting RhoA- or Rac1-dependent functions. Short-term ZCL278 exposure in primary murine neonatal cortical neurons is shown to dramatically abolish neurite branching (50μM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
Description 94% by HPLC
Target Proteins CDC42
Molecular Formula C21H19BrClN5O4S2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.