Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cdk7 Inhibitor VIII, THZ1

Catalog No. 5323720001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-237-20001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-237-20001 Supplier MilliporeSigma™ Supplier No. 5323720001
Add to Cart
Add to Cart

A cell-permeable, non-toxic, phenylaminopyrimidinyl-acrylamide compound that acts as a highly potent, selective, ATP-site directed, and irreversible inhibitor of Cdk7 (IC50

A cell-permeable, non-toxic, phenylaminopyrimidinyl-acrylamide compound that acts as a highly potent, selective, ATP-site directed, and irreversible inhibitor of Cdk7 (IC50 = 3.2nM). Acts by allosterically and covalently modifying Cys312 located outside the canonical kinase domain. Shown to block the phosphorylation of Ser5 and Ser7 in RNAPII CTD, the intracellular substrate of Cdk7 (< 250 nM, 4 h in Jurkat cells). Does not affect the activity of C312S mutated Cdk7. Exhibits comparatively week activity towards Cdk12 (IC50 = 250nM), Cdk1, and Cdk2. Suppresses the proliferation and cell viability of T-ALL and CLL cell lines and induces apoptosis with significant reduction in the levels of MCL-1 and XIAP. Also induces a significant reduction in the transcript and protein levels of RUNX1, TAL, and GATA3 (∽ 50nM). Suppresses the growth of KOPTK1 cells xenografts in murine model (10mg/kg, b.i.d).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 96% by HPLC
Target Proteins Cdk7
Molecular Formula C31H28ClN7O2·4HCl
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.