Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CFTR Activator, Cact-A1

Catalog No. 5059850001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-598-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-598-50001 Supplier MilliporeSigma™ Supplier No. 5059850001
Add to Cart
Add to Cart

A cell-permeable, non-toxic aminopyrazolocarbonitrile compound that acts as a potent, selective, reversible and cAMP-independent activator of CFTR-dependent Cl- channel in airway epithelial cells (EC50

A cell-permeable, non-toxic aminopyrazolocarbonitrile compound that acts as a potent, selective, reversible and cAMP-independent activator of CFTR-dependent Cl- channel in airway epithelial cells (EC50 = 1.6μM in CFTR-expressing Fisher rat thyroid cells). Also shown to activate δF508-CFTR in primary CF-HBE cell cultures (EC50 = 3.5μM). Reported to directly bind to the same site as CFTRinh-172 ( Cat. No. 219670, Cat. No. 219674). Does neither elevate intracellular cAMP levels nor appear to have any requirement for cAMP agonist, such as forskolin (Cat. No. 344270, Cat. No. 344282). Shown to additively potentiate the G551D-CFTR Cl- current activated by forskolin with CFTR potentiator, VX-770. Exerts no effect on either Ca2+-activated Cl- channel (TMEM16A) or intracellular Ca2+ levels.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Description 98% by HPLC
Target Proteins CFTR
Molecular Formula C16H13N5O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.