Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CFTR Inhibitor IV, PPQ-102

Catalog No. 21967710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-967-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-967-710MG Supplier MilliporeSigma™ Supplier No. 21967710MG
Add to Cart
Add to Cart

The CFTR Inhibitor IV, PPQ-102, also referenced under CAS 931706-15-9, controls the biological activity of CFTR. This small molecule/inhibitor is primarily used for Neuroscience applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
CAS 931706-15-9
Molecular Formula C26H22N4O3
Formula Weight 438.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Crystalline Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.