Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CFTR Potentiator, VX-770

Catalog No. 5305410001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-054-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-054-10001 Supplier MilliporeSigma™ Supplier No. 5305410001
Add to Cart
Add to Cart

A cell-permeable, potentiator of CFTR that directly, reversibly, and allosterically binds to WT and G551D mutant forms (≈10 μM) and increases the open probability to restore lung function.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins WT and G551D mutant forms of cell surface CFTR
CAS 873054-44-5
Molecular Formula C24H28N2O3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.