Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CGP 7930

Catalog No. 5058940001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-589-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-589-40001 Supplier MilliporeSigma™ Supplier No. 5058940001
Add to Cart
Add to Cart

A blood brain barrier permeable, positive allosteric modulator of the metabotropic GABAB recep

A blood brain barrier permeable, positive allosteric modulator of the metabotropic GABAB receptor. Shown to modulate GABAB receptor in the high affinity state and thereby increases agonist affinity for the receptor. Also enhances agonist potency and efficacy even on recombinant GABAB receptors. Exhibits anxiolytic effects in rodents.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins GABAB
Molecular Formula C19H32O2
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.