Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Chk2 Inhibitor II

Catalog No. 2204861MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-048-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-048-61MG Supplier MilliporeSigma™ Supplier No. 2204861MG
Add to Cart
Add to Cart

Chk2 Inhibitor II, BML-277, CAS 516480-79-8 is a cell-permeable, potent, reversible, ATP-competitive inhibitor of Chk2 (IC50 = 15 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 95% by HPLC
Target Proteins Chk2
CAS 516480-79-8
Molecular Formula C20H14ClN3O2
Formula Weight 363.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.