Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cinnabarinic acid

Catalog No. 5048200001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-482-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-482-00001 Supplier MilliporeSigma™ Supplier No. 5048200001
Only null left
Add to Cart
Add to Cart

A partial, low-potency agonist highly selective for mGlu4 receptors (EC50 > 100μM, no effect on other mGlu subtype rece

A partial, low-potency agonist highly selective for mGlu4 receptors (EC50 > 100μM, no effect on other mGlu subtype receptors). Used in assessing the functional roles of mGlu4 receptors in a variety of research areas, such as excitotoxic cell death, apoptosis, neuroinflammation, inflammation, and neuroimmune communication.
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description 98% by HPLC
Target Proteins mGlu4 receptors
Molecular Formula C14H8N2O6
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.