Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Clioquinol

Catalog No. 2331651GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-316-51GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-316-51GM Supplier MilliporeSigma™ Supplier No. 2331651GM
Add to Cart
Add to Cart

A metal ion chelator that crosses the blood brain barrier and acts as a neurotoxic antibiotic.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins Metal ion chelator
CAS 130-26-7
Molecular Formula C9H5ClINO
Formula Weight 305.5g/mol
Solubility DMSO, EtOH or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.