Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Clodronate, Disodium Salt

Catalog No. 23318310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-318-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-318-310MG Supplier MilliporeSigma™ Supplier No. 23318310MG
Add to Cart
Add to Cart

A non-amino, chloro-containing bisphosphonate that acts as a potent inhibitor of osteoclast-mediated bone resorption.

A non-amino, chloro-containing bisphosphonate that acts as a potent inhibitor of osteoclast-mediated bone resorption. It is metabolized into a non-hydrolyzable ATP analog in osteoclasts and macrophages, and induces apoptosis by inhibiting mitochondrial function. An efficient metal chelator that inhibits invasion, adhesion, and migration of PC-3 prostate cancer cells (IC50 = 100 nM).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by titration
Target Proteins Osteoclast-mediated bone resorption
CAS 22560-50-5
Molecular Formula CH2Cl2O6P2Na2·4H2O
Formula Weight 360.9g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.