Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (+/-)-Clopidogrel Hydrogensulfate

Catalog No. 5057620001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-576-20001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-576-20001 Supplier MilliporeSigma™ Supplier No. 5057620001
Add to Cart
Add to Cart

A thienopyridine compound that acts as a specific irreversible inhibitor of purinergic P2Y12 recept

A thienopyridine compound that acts as a specific irreversible inhibitor of purinergic P2Y12 receptors. Blocks ADP-induced platelet aggregation and serves as an anti-thrombotic agent. Shown to reduce vascular inflammation and angiotensin II induced-abdominal aortic aneurysm progression. Clopidogrel is a prodrug that requires activation by CYP enzyme system to produce its pharmacologically active metabolite.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins P2Y12
Molecular Formula C16H16ClNO2S·H2SO4
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.