Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CNQX

Catalog No. 5049140001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-491-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-491-40001 Supplier MilliporeSigma™ Supplier No. 5049140001
Add to Cart
Add to Cart

A potent and competitive antagonist for AMPA/kainate receptors (Ki

A potent and competitive antagonist for AMPA/kainate receptors (Ki = 0.6 - 2.11μM for GluA1-4 and Ki = 1.3-5.3μM for different subtypes of Kainate receptors) and a negative allosteric modulator at NMDA receptor glycine binding site. Widely used for assessing the functional roles of neurotransmission mediated by glutamate receptors in both normal and abnormal brain functions.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Cream
Description 99% by HPLC
Target Proteins AMPA
Molecular Formula C9H4N4O4
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.