Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Coenzyme A, Trilithium Salt

Catalog No. 234101100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-410-1100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-410-1100MG Supplier MilliporeSigma™ Supplier No. 234101100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serves as cofactor in enzymatic acetyl transfer reactions

  • Hygroscopic in nature
  • Assay: ≥94% by UV
  • CAS: 18439-24-2
  • Molecular Formula: C21H33N7O16P3S · 3Li
  • Color: Off-White
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.