Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Colcemid

Catalog No. 2341095MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-410-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-410-95MG Supplier MilliporeSigma™ Supplier No. 2341095MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell synchronization agent.

Cell synchronization agent. Induces apoptosis by blocking mitosis in HeLa S3 cells. Depolymerizes microtubules and limits microtubule formation. Appropriate dilutions inactivate spindle fiber mechanism during metaphase, systematically immobilizing chromosomes at the metaphase plate.
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins Depolymerizes microtubules and limits microtubule formation.
CAS 477-30-5
Molecular Formula C21H25NO5
Formula Weight 371.4g/mol
Solubility EtOH
Form Crystalline Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.