Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Collagenase Inhibitor I

Catalog No. 2341405MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-414-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-414-05MG Supplier MilliporeSigma™ Supplier No. 2341405MG
Add to Cart
Add to Cart

The Collagenase Inhibitor I controls the biological activity of Collagenase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: Z-Pro-D-Leu-D-Ala-NHOH
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by TLC
Target Proteins vertebrate collegenases
Molecular Formula C22H32N4O6
Formula Weight 448.5g/mol
Solubility EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.