Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Combretastatin A-4

Catalog No. 23420010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-420-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-420-010MG Supplier MilliporeSigma™ Supplier No. 23420010MG
Add to Cart
Add to Cart

A synthetic Combretum caffrum-derived cell-permeable stilbenoid phenol that competes with colchicine tubulin binding and interferes with tubulin polymerization as well as cellular microtubule assembly

A synthetic Combretum caffrum -derived cell-permeable stilbenoid phenol that competes with colchicine (Cat No. 234115) tubulin binding (by 96.4% at equal molar concentration; Ki = 1.1μM) and interferes with tubulin polymerization (IC50 = 2-3μM) as well as cellular microtubule assembly. In addition to its antimitotic and in vitro cancer growth inhibition activity (IC50 = 7nM in L1210 cultures), CA4 also acts as an antivascular agent and effectively induces tumor cell necrosis in vivo by targeting endothelial cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 117048-59-6
Molecular Formula C18H20O5
Formula Weight 316.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.