Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ω-Conotoxin GVIA, Conus geographus

Catalog No. 343781100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-378-1100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-378-1100UG Supplier MilliporeSigma™ Supplier No. 343781100UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A neurotoxin that acts as an antagonist of voltage-activated N-type Ca2+ channels and of neurotransmitter release at neuronal synapses.

Peptide Sequence: H-Cys1-Lys-Ser-Hyp-Gly-Ser-Ser-Cys8-Ser-Hyp-Thr-Ser-Tyr-Asn-Cys15-Cys16-Arg-Ser-Cys19-Asn-Hyp-Tyr-Thr-Lys-Arg-Cys26-Tyr-NH2 (disulfide bonds: 1 → 16; 8 → 19; 15 → 26)
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins Voltage-activated N-type Ca2+ channels
CAS 106375-28-4
Molecular Formula C120H182N38O43S6
Formula Weight 3037.4g/mol
Solubility Aqueous Buffers or H2O
Form Lyophilized
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.