Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ COX-1 Inhibitor, FR122047

Catalog No. 2360055MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-600-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-600-55MG Supplier MilliporeSigma™ Supplier No. 2360055MG
Add to Cart
Add to Cart

The COX-1 Inhibitor, FR122047, also referenced under CAS 130717-51-0, controls the biological activity of COX-1. This small molecule/inhibitor is primarily used for Cell Signaling applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins COX-1, COX-2
CAS 130717-51-0
Molecular Formula C23H25N3O3S·HCl
Formula Weight 460g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.