Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ cPLA2α Inhibitor II, Pyrrophenone

Catalog No. 5305380001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-053-80001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-053-80001 Supplier MilliporeSigma™ Supplier No. 5305380001
Add to Cart
Add to Cart

A cell-permeable, highly potent, and reversible inhibitor of cytosolic phospholipase A2a (IC50 = 4.2 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins PLA2a
Molecular Formula C49H37F2N3O5S2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.