Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CTAP

Catalog No. 5083160001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-831-60001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-831-60001 Supplier MilliporeSigma™ Supplier No. 5083160001
Add to Cart
Add to Cart

A highly potent octapeptide that acts as a selective and brain penetrating bioavailableμ opioid receptor antagonist (IC50

A highly potent octapeptide that acts as a selective and brain penetrating bioavailableμ opioid receptor antagonist (IC50 = 3.5nM). Used in studies of reward systems and additions.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins u Opioid receptpr
Molecular Formula C51H69N13O11S2
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.