Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CTOP

Catalog No. 5080150001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-801-50001 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-801-50001 Supplier MilliporeSigma™ Supplier No. 5080150001
Add to Cart
Add to Cart

A potent antagonist highly selective forμ opioid receptor (Ki

A potent antagonist highly selective forμ opioid receptor (Ki = 0.96nM forμ receptor and > 10,000nM for δ receptors). Often used for studying the functional roles ofμ opioid receptors in both central and peripheral nervous systems.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as trifluoracetate salt
Description 98% by HPLC
Target Proteins mu opioid
Molecular Formula C50H67N11O11S2
Formula Weight 1062.28g/mol
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.