Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ CXCR7 Agonist, VUF11207

Catalog No. 23982410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-982-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-982-410MG Supplier MilliporeSigma™ Supplier No. 23982410MG
Add to Cart
Add to Cart

The CXCR7 Agonist, VUF11207 controls the biological activity of CXCR7. This small molecule/inhibitor is primarily used for Biochemicals applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Description 99% by HPLC
Target Proteins CXCR7
Formula Weight 470.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Oil
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.