Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cyclo(Arg-Gly-Asp-D-Phe-Val)

Catalog No. 1820151MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-201-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-201-51MG Supplier MilliporeSigma™ Supplier No. 1820151MG
Add to Cart
Add to Cart

Potent inhibitor of cell adhesion.

Potent inhibitor of cell adhesion. Inhibits tumor cell adhesion to laminin and vitronectin substrates. In vivo investigations in rats showed it to be useful in the amelioration of ischemic acute renal failure in rats. Also inhibits tubular obstruction by preventing cell-cell adhesion. Inhibits the redistribution of c-Src into focal adhesions, thereby leading to impaired MAP Kinase activation.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt
Description 95% by HPLC
Target Proteins inhibits tumor cell adhesion to laminin and vitronectin substrates
CAS 137813-35-5
Molecular Formula C26H38N8O7
Formula Weight 574.6g/mol
Solubility 1N Acetic Acid
Content And Storage Hygroscopic
Form Solid
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.