Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cyclopamine-KAAD

Catalog No. 239804500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-980-4500UG 500 μg
23-980-4100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-980-4500UG Supplier MilliporeSigma™ Supplier No. 239804500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopamine- KAAD, CAS 306387-90-6, is a cell-permeable potent analog of Cyclopamine (Cat. No. 239803) that specifically inhibits Hedgehog (Hh) signaling with similar or lower toxicity (IC50 = 20 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Target Proteins Hh signaling in Shh-light2 assay
Molecular Formula C44H63N3O4
Formula Weight 698g/mol
Solubility DMSO, EtOH, or MeOH
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Purity 70% by HPLC (sum of two isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.