Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cyclophosphamide Monohydrate

Catalog No. 2397851GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-978-51GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-978-51GM Supplier MilliporeSigma™ Supplier No. 2397851GM
Add to Cart
Add to Cart

A widely used anti-neoplastic agent that also displays immunosuppressant properties.

A widely used anti-neoplastic agent that also displays immunosuppressant properties. Induces apoptotic cell death in rat thymus through the Fas/Fas ligand pathway.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Fas/Fas ligand pathway
CAS 6055-19-2
Molecular Formula C7H15Cl2N2O2P·H2O
Formula Weight 279.1g/mol
Solubility H2O
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.